Immunoassays feature
Choose high-quality Invitrogen immunoassays for protein quantitation
Rely on our 30 years of experience manufacturing and supplying antibody-based, target-specific protein quantitation reagents, kits, and panels for life science research. These products, which are collectively called immunoassays, include matched antibody pairs, uncoated and coated-plate ELISA kits, and multiplexing assays for the Luminex platform. All are validated for reliability and specificity. Select from hundreds of ready-to-ship kits or configure and order a custom assay panel.
To find an exceptional assay for your needs, choose from popular immunoassay categories below or contact our field application specialists (FAS), who can answer your questions and provide you with individualized immunoassay support, including information about new immunoassay technologies.
NEW ProcartaPlex Signaling Dual Reporter assay
Revolutionize your way to unveil the complex interactions in studying cell signaling pathways. Effortlessly detect and quantify total and phosphorylated signaling proteins simultaneously in a single well.
Learn more
Featured products
Gene expression directly from a lysate
Multiplex 80 RNA gene expression targets directly from a lysate in a single well.
Learn more
Enter the next dimension of protein multiplexing
Combine up to 80 protein targets from complex biological samples, with flexible options from preconfigured solutions to custom mixed panels.
Learn more
Which immunoassay platform are you looking for?
ProQuantum high-sensitivity immunoassays
Invitrogen ProQuantum immunoassays offer high sensitivity and broad dynamic range, using only 2–5 µL of sample. The simple workflow requires no wash steps and is especially convenient for qPCR users, as these immunoassays run on any qPCR instrument and do not require any specialized instruments.
Order now
ProQuantum immunoassay features to make the best choice for your current project
Multiplexable
No
Sample volume used
2–5 μL
Best used for
Validation
Catalog size
1 x 96 reactions,
5 x 96 reactions
Dynamic range
≥5 logarithmic units
Analytical sensitivity
femtogram/mL
Sample-to-result time
2 hr
Multiplexable
Yes
Sample volume used
25 μL
Best used for
Screening
Catalog size
1 plate (96 tests)
Dynamic range
3–4 logarithmic units
Analytical sensitivity
picogram/mL
Sample-to-result time
4.5 hr
Multiplexable
No
Sample volume used
50 μL
Best used for
Validation
Catalog size
1 plate (96 tests),
2 plates, 5 plates
Dynamic range
2–3 logarithmic units
Analytical sensitivity
picogram/mL
Sample-to-result time
4 hr
Multiplexable
No
Sample volume used
50 μL
Best used for
Validation
Catalog size
2 plates,
10 plates, 20 plates
Dynamic range
2–3 logarithmic units
Analytical sensitivity
picogram/mL
Sample-to-result time
24 hr
Multiplexable
No
Sample volume used
50 μL
Best used for
Validation
Catalog size
N/A
Dynamic range
N/A
Analytical sensitivity
N/A
Sample-to-result time
N/A
ProQuantum
immunoassays
Multiplex
immunoassays
Coated
ELISA kits
Uncoated
ELISA kits
Build-it-yourself
immunoassays
Learn more
Learn more
Learn more
Learn more
Learn more
Inside cell11 luminex eyecandy
Immunoassay Research Areas
Explore our immunoassays for different research areas, including immune-oncology, neurobiology, isotyping and immunoglobulin characterization, hallmarks of cancer, car t cells, and more.
Learn more
Starting a new research project? We offer a variety of services to help.
Shop antibodies
Select antibodies to build your own ELISA
We offer a variety of specific antibodies to help you.
Shop antibodies
Custom ELISA or ProQuantum immunoassay
Request bulk packaging, lot reservation, or go custom
Contact us and we can help you with a variety of special requests to help you with the start of your next project.
Request a quote
Illustration of SARS-CoV-2 virus
SARS-CoV-2 Serology Assays
Rapidly detect antibodies (immunoglobulins, IgG and IgM) in serum against the SARS-CoV-2 coronavirus.