Correlative Electron Microscopy Software
Thermo Scientific Maps Software is an imaging and correlative workflow software suite compatible with the full line of Thermo Scientific SEM, DualBeam (FIB SEM) and TEM platforms.
Scientists and researchers rely increasingly on nanoscale observations to inform the latest advances in research and analysis. It has, however, become apparent that high-resolution observations lose much of their utility in the absence of the larger macroscopic context. Observations from multiple sources must be linked, providing the necessary multi-scale and multi-modal insight for truly valuable data.
Maps Software provides a powerful imaging workflow automation package within an easy-to-use and robust platform. With just a few clicks, you can collect impactful data while preserving the context of your observations.
Multi-scale imaging automation
Whether its scanning electron microscopy analysis in SEM and DualBeam tools or transmission electron microscopy (TEM), Maps Software enables automated 2D imaging for a variety of applications, from acquiring large image mosaics to scheduling routine imaging tasks for off times (overnight, weekends).
  • Acquire up to four simultaneous signals, automatically (STEM/SEM)
  • Run multiple samples in series to increase system productivity
  • Choose rectangular, circular or custom shapes for mapping acquisition (STEM/SEM)
Diatom identified in sand with multi scale microscopy.
Diatom identification in sand with multi-scale analysis in Maps Software.
Correlative microscopy
With the ability to correlate optical and electron microscope data and easy navigation from SEM to TEM, Maps Software allows you to obtain the necessary context that drives your research and development. Use multi-modal data to aid in interpretation and navigation, ensuring you are always collecting the right data from the right location.
  • Import 2D and 3D data from any source
  • Quickly generate context for the entire sample grid or targeted region of interest
  • Register imagery from other modalities (e.g. energy-dispersive spectroscopy, secondary electron signal, electron backscatter diffraction, etc.)
  • Integrate observations in a multi-scale, multi-layered visualization environment
  • Align data with system stages using any Thermo Scientific EM platform
Correlative microscopy of an Inconel 718 part, combining optical and electron microscopy imaging with EBSD analysis.
Correlative microscopy of an Inconel 718 part, combining optical and electron microscopy imaging with EBSD analysis.
Integrated analytics
Maps Software supports integrated EDS map acquisition on STEM platforms for chemical characterization and elemental analysis along with your high-resolution imaging.
  • Automate large-area, high-throughput EDS acquisition with the Thermo Scientific Super-X and Dual-X Detector Systems
  • Enable native correlation with electron imaging layers
  • Employ built-in stitching and multiscale visualization
  • Easily view EDS maps away from the microscope with the optional offline version of Maps Software
Intuitive visualization and offline collaboration
The optional offline version of Maps Software lets you take the microscope with you when you are away from the lab.
  • View your data from your own PC
  • Access the full correlative power of Maps Software anywhere
  • Plan for your next imaging session before you head to the lab
  • Easily share your multi-scale observations with colleagues
Avizo Software integration
The addition of Thermo Scientific Avizo Software automates your image processing and statistical data generation. Avizo Software provides a rich, integrated user experience that blends acquisition with advanced analysis, and works in the background to process images as they are acquired.
  • Gain instant access to critical statistical data through a complete, automated imaging and analysis solution
  • Perform advanced image processing, accessible while you are on the tool
  • Tune and optimize imaging and processing workflows
Gold nanoparticle analysis performed with Avizo Software.
Avizo Software provides automated nanoparticle analysis in TEM. Here, EDS analysis counted the number of particles containing chlorine (light blue) nickel (green) and gold (red) and produced a histogram of their relative particle sizes. Sample courtesy of J. Bursik, Institute of Physics of Materials, Brno.
Key Features
  • Compatibility with current Thermo Scientific SEM, FIB-SEM, and TEM platforms; providing a unified user experience across all instrument types.
  • Integrated approach to data acquisition, annotation and storage.
  • Automated acquisition of large overviews at any magnification (tile and stitch); multiple acquisitions can be set up for unsupervised batch data collection.
  • Setup and coordination of experiments across multiple microscopes.
  • Correlation of images from different microscopes, including imported images from 3rd party instruments using the integrated Bio-Formats Library.
Multi-scale imaging automation
A completely automated solution for generating high-quality, multi-scale images.
Correlative microscopy
Gain insight by integrating observations across modalities and imaging platforms.
Integrated analytics
Make EDS mapping easier and instantly valuable via direct acquisition and integration of elemental layers.
Intuitive visualization and offline collaboration
The optional offline version of Maps Software allows you to continue your investigation and plan your next imaging session while away from the microscope.
Maps offline viewer
This free, standalone application allows you to view Maps project from your PC (Windows 7 or 10). After installation, simply launch the application and import your projects for viewing.
Maps Offline Viewer allows you to open Maps projects on any PC. It also allows easier sharing of results without requiring a licensed version of Maps. Note that the viewer does not, however, allow processing steps like stitching, etc.
Download Maps offline viewer v3 ›
For users with images created using Maps version 3.0 or higher
Download Maps offline viewer v2 ›
For users with images created using Maps version 2.5 or lower
Style Sheet for Products Table Specifications
Style Sheet for Komodo Tabs
Resources
Video Player is loading.
Current Time 0:00
Duration 2:25
Loaded: 6.77%
Stream Type LIVE
Remaining Time 2:25
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Webinar: Boost Productivity in Multi-Scale and Multi-Modal Material Analysis Challenges
    In this webinar, you will:
    • Discover the beauty of high-resolution images in a larger context.
    • Learn how Maps Software efficiently handles a large number of samples at the same time.
    • See how data from different sources is combined to answer questions no individual technique can answer.
    • Learn how Maps Software handles navigation, alignment and correlation in practice.
    • Understand how sharp images can be acquired without an operator being present.
    • Find out how Maps Software can function as a powerful “virtual SEM”.
    • Discover what sharing images and data looks like to your collaborators.
    Video Player is loading.
    Current Time 0:00
    Duration 2:25
    Loaded: 3.44%
    Stream Type LIVE
    Remaining Time 2:25
    1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
      • en (Main), selected
      Webinar: Boost Productivity in Multi-Scale and Multi-Modal Material Analysis Challenges
      In this webinar, you will:
      • Discover the beauty of high-resolution images in a larger context.
      • Learn how Maps Software efficiently handles a large number of samples at the same time.
      • See how data from different sources is combined to answer questions no individual technique can answer.
      • Learn how Maps Software handles navigation, alignment and correlation in practice.
      • Understand how sharp images can be acquired without an operator being present.
      • Find out how Maps Software can function as a powerful “virtual SEM”.
      • Discover what sharing images and data looks like to your collaborators.
      Applications
      Infectious disease research
      Infectious Disease Research
      Cryo-EM techniques enable multiscale observations of 3D biological structures in their near-native states, informing faster, more efficient development of therapeutics.
      Drug discovery
      Drug Discovery
      Learn how to take advantage of rational drug design for many major drug target classes, leading to best-in-class drugs.
      Plant biology research
      Plant Biology Research
      Fundamental plant biology research is enabled by cryo electron microscopy, which provides information on proteins (with single particle analysis), to their cellular context (with tomography), all the way up to the overall structure of the plant (large volume analysis).
      Pathology research
      Pathology Research
      Transmission electron microscopy (TEM) is used when the nature of the disease cannot be established via alternative methods. With nano-biological imaging, TEM provides accurate and reliable insight for certain pathologies.
      Process control using electron microscopy
      Process control using electron microscopy
      Modern industry demands high throughput with superior quality, a balance that is maintained through robust process control. SEM and TEM tools with dedicated automation software provide rapid, multi-scale information for process monitoring and improvement.
      Quality control and failure analysis using electron microscopy
      Quality control and failure analysis
      Quality control and assurance are essential in modern industry. We offer a range of EM and spectroscopy tools for multi-scale and multi-modal analysis of defects, allowing you to make reliable and informed decisions for process control and improvement.
      Fundamental Materials Research_R&D_Thumb_274x180_144DPI
      Fundamental Materials Research
      Novel materials are investigated at increasingly smaller scales for maximum control of their physical and chemical properties. Electron microscopy provides researchers with key insight into a wide variety of material characteristics at the micro- to nano-scale.
      Semiconductor-RD_1920x1080
      Semiconductor research and development
      Innovation starts with research and development. Learn more about solutions to help you understand innovative structures and materials at the atomic level.
      Semi-Metrology_1920x1080
      Semiconductor metrology
      Manufacturing today’s complex semiconductors requires exact process controls. Learn more about advanced metrology and analysis solutions to accelerate yield learnings.
      Semiconductor Failure Analysis
      Semiconductor Failure Analysis
      Complex semiconductor device structures result in more places for defects to hide. Learn more about failure analysis solutions to isolate, analyze, and repair defects.
      Semi-Materials-Character_1920x1080
      Semiconductor materials characterization
      Many factors impact yield, performance, and reliability. Learn more about solutions to characterize physical, structural, and chemical properties.
      Techniques
      Single Particle Analysis
      Single particle analysis (SPA) is a cryo-electron microscopy technique that enables structural characterization at near-atomic resolutions, unraveling dynamic biological processes and the structure of biomolecular complexes/assemblies.
      Learn more ›
      Cryo-Tomography
      Cryo-electron tomography (cryo-ET) delivers both structural information about individual proteins as well as their spatial arrangements within the cell. This makes it a truly unique technique and also explains why the method has such an enormous potential for cell biology. Cryo-ET can bridge the gap between light microscopy and near-atomic-resolution techniques like single-particle analysis.
      Learn more ›
      MicroED
      MicroED is an exciting new technique with applications in the structural determination of small molecules and protein. With this method, atomic details can be extracted from individual nanocrystals (<200 nm in size), even in a heterogeneous mixture.
      Learn more ›
      Particle analysis
      Particle analysis plays a vital role in nanomaterials research and quality control. The nanometer-scale resolution and superior imaging of electron microscopy can be combined with specialized software for rapid characterization of powders and particles.
      Learn more ›
      3D EDS Tomography
      Modern materials research is increasingly reliant on nanoscale analysis in three dimensions. 3D characterization, including compositional data for full chemical and structural context, is possible with 3D EM and energy dispersive X-ray spectroscopy.
      Learn more ›
      EDS Elemental Analysis
      Thermo Scientific Phenom Elemental Mapping Software provides fast and reliable information on the distribution of chemical elements within a sample.
      Learn more ›
      Semiconductor TEM Imaging and Analysis
      Thermo Scientific transmission electron microscopes offer high-resolution imaging and analysis of semiconductor devices, enabling manufacturers to calibrate toolsets, diagnose failure mechanisms, and optimize overall process yields.
      Learn more ›
      Semiconductor Analysis and Imaging
      Thermo Fisher Scientific offers scanning electron microscopes for every function of a semiconductor lab, from general imaging tasks to advanced failure analysis techniques requiring precise voltage-contrast measurements.
      Learn more ›
      Automated Particle Workflow
      The Automated NanoParticle Workflow (APW) is a transmission electron microscope workflow for nanoparticle analysis, offering large area, high resolution imaging and data acquisition at the nanoscale, with on-the-fly processing.
      Learn more ›
      Single Particle Analysis
      Single particle analysis (SPA) is a cryo-electron microscopy technique that enables structural characterization at near-atomic resolutions, unraveling dynamic biological processes and the structure of biomolecular complexes/assemblies.
      Learn more ›
      Cryo-Tomography
      Cryo-electron tomography (cryo-ET) delivers both structural information about individual proteins as well as their spatial arrangements within the cell. This makes it a truly unique technique and also explains why the method has such an enormous potential for cell biology. Cryo-ET can bridge the gap between light microscopy and near-atomic-resolution techniques like single-particle analysis.
      Learn more ›
      MicroED
      MicroED is an exciting new technique with applications in the structural determination of small molecules and protein. With this method, atomic details can be extracted from individual nanocrystals (<200 nm in size), even in a heterogeneous mixture.
      Learn more ›
      Particle analysis
      Particle analysis plays a vital role in nanomaterials research and quality control. The nanometer-scale resolution and superior imaging of electron microscopy can be combined with specialized software for rapid characterization of powders and particles.
      Learn more ›
      3D EDS Tomography
      Modern materials research is increasingly reliant on nanoscale analysis in three dimensions. 3D characterization, including compositional data for full chemical and structural context, is possible with 3D EM and energy dispersive X-ray spectroscopy.
      Learn more ›
      EDS Elemental Analysis
      Thermo Scientific Phenom Elemental Mapping Software provides fast and reliable information on the distribution of chemical elements within a sample.
      Learn more ›
      Semiconductor TEM Imaging and Analysis
      Thermo Scientific transmission electron microscopes offer high-resolution imaging and analysis of semiconductor devices, enabling manufacturers to calibrate toolsets, diagnose failure mechanisms, and optimize overall process yields.
      Learn more ›
      Semiconductor Analysis and Imaging
      Thermo Fisher Scientific offers scanning electron microscopes for every function of a semiconductor lab, from general imaging tasks to advanced failure analysis techniques requiring precise voltage-contrast measurements.
      Learn more ›
      Automated Particle Workflow
      The Automated NanoParticle Workflow (APW) is a transmission electron microscope workflow for nanoparticle analysis, offering large area, high resolution imaging and data acquisition at the nanoscale, with on-the-fly processing.
      Learn more ›
      Style Sheet to change H2 style to p with em-h2-header class
      Contact us
      Electron microscopy services
      To ensure optimal system performance, we provide you access to a world-class network of field service experts, technical support, and certified spare parts.
      Learn more ›
      Style Sheet for Support and Service footer
      Style Sheet for Fonts
      Style Sheet for Cards