mid-polar-showcase-image-230x195
Designed to deliver robust performance for specific applications
We designed these Thermo Scientific GC columns to exceed the methodology demands of specific applications. They deliver robust performance, high levels of reproducibility, superior inertness, and sensitivity, coupled with fast analysis times.
Contact us
Application specific gas chromatography columns
Common applications:
  • PAH and PBDE
  • Dioxin/Furan and PCB
  • Organophosphorus and organochlorine pesticides
  • Very volatile (VVOC) and volatile organic compounds (VOC)
  • Amphetamines and other drugs of abuse
  • Volatile and fatty acid components of biodiesel
ColumnRecommended Application
TraceGOLD TG-PAH GC ColumnsUse any of those thermally stable GC columns for fast analysis of regulated PAHs.
TraceGOLD TG-PBDE GC ColumnsFast analysis of PBDE with high BDE-209 response, ideal for U.S. EPA method 1614.
TraceGOLD TG-XLBMS GC ColumnsUse these low–polarity, low–bleed GC Columns for analyses of pesticides, PCB congeners, and polyaromatic hydrocarbons (PAHs).
TraceGOLD TG-VRX GC ColumnsAnalyze volatile organic pollutants and aromatics with these GC Columns that provide good resolution and fast analysis times and are ideally suited for US EPA Methods 502.2, 524.2, 8021, and 8260.
TraceGOLD TG-VMS GC ColumnsAchieve good resolution and fast analysis times for volatile organic pollutants with TraceGOLD TG-VMS GC Columns.
TraceGOLD TG-OCP I and II GC ColumnsAnalyze organochlorine pesticides and herbicides with these ultra low bleed, highly sensitive columns that provide excellent peak shape and the highest levels of reproducibility; proprietary formulation facilitates analysis of difficult compounds.
TraceGOLD TG-OPP I and II GC ColumnsAnalyze organophosphorus pesticides with ultra-low bleed, superior inertness and consistent reproducibility; ideal for U.S. EPA methods 8141A.
TraceGOLD TG-Dioxin GC ColumnsUse any of these thermally stable GC columns as a confirmation GC column with unique selectivity for toxic dioxin and furan congeners to achieve unsurpassed performance and reproducibility.
TRACE GC Columns for Dioxin and PCB AnalysisCustom–tailored low–polarity GC Columns for dioxin and PCB Analysis optimized to deliver unsurpassed performance and reproducibility and provide wide coverage of the dioxin and PCB congeners with the highest toxicological significance.
TRACE GC Columns for Specific EPA MethodsPerform specific EPA methodologies with confidence and unsurpassed performance using these specially designed capillary GC columns.
TRACE GC Columns for PesticidesIdentify organophosphorus and organochlorine pesticides using GC columns that meet the most stringent requirements for pesticide and herbicide analysis.
TRACE TR-BioDiesel GC ColumnsDetermine methanol, fatty acid methyl esters (FAMEs) and glycerides in biodiesel analysis with high performance and reproducibility.
TRACE GC Columns for Drugs of AbuseAnalyze amphetamines, codeine, morphine, and THC with these GC columns designed for high performance, sensitivity and reproducibility.
TraceGOLD TG-VVOC BAchieve excellent peak shapes and great stability for very volatile compounds including volatile amines, ozone precursors and air toxics.
Chromatography Consumables Resources and Information
Find interactive tools and calculators, guides to select the right products, equivalent product finders, and get in contact with our consumables technical support team to answer your application questions.
Discover more resources
Need to find GC columns or accessories?
Access the latest interactive version of our Chromatography Consumables & Columns Catalog. Our enhanced online version features easier access to product information, key application notes and brand-new selector tools.
Get the GC columns catalog
CMD SchemaApp code