Multiplexing instrumentation for RNA and protein quantitation
The Luminex family of systems encompasses four instrument models, each offering distinct multiplexing capabilities, throughput, and read times. These Luminex instruments operate on the quantitative fluorescent flow cytometry principle, leveraging the xMAP magnetic bead technology to detect small quantities of analytes. Used in combination with our ProcartaPlex multiplex assays for protein quantitation or QuantiGene Plex assays for gene expression analysis, the Luminex platform delivers a rapid and cost-effective immunoassay profiling tool. This synergy between Luminex instruments and our advanced multiplex assays enhances efficiency and precision in the detection and quantification of biomolecules.
Which Luminex model is right for you?
  xMAP INTELLIFLEX 
DR-SE
 System
xMAP INTELLIFLEX 
System
FLEXMAP 3D
System
Luminex 200
System
  Double the data with the second reporter channel Compact footprint with our broadest dynamic range Fast read-time and high-throughput Fast and robust platform for up to 100 analytes
Performance
Multiplex capacity
Up to 500
Up to 2 parameters per analytes
Up to 500 Up to 500
Up to 100
(80 on MagPlex)
Dynamic range
≥ 5.5 logs (RP1)
≥ 4.5 logs (RP2)
≥ 5.5 logs (RP1)
≥ 4.5 logs (RP1)
≥ 3.5 logs (RP1)
Read time
96-well in ~20 min
384-well in ~75 min
96-well in ~20 min
384-well in ~75 min
96-well in ~20 min
384-well in ~75 min
45 min (96-well)
Sensitivity (assay dependent)
0.06–1 pg/mL (Protein assays); 1,000 RNA copies per target/well (RNA assays)
Technical
Footprint including PC (linear bench space)
58.4 cm x 61 cm x 76 cm
58.4 cm x 61 cm x 76 cm
110 cm x 62 cm x 63 cm
115 cm x 60 cm x 50 cm
Weight
54.5 kg (120 lb)
54.5 kg (120 lb)
77.1 kg (170 lb)
49 kg (113 lb)
Optics
Flow cytometry-based lasers, APDs, PMTs
Reporter laser/optics
532 nm (green) and 405 nm (violet)
532 nm (green) 
532 nm (green)
532 nm (green)
Automation-compatible hardware
Front and side eject
Front eject only
Front eject only
Front eject only
Operational validation
Calibration, verification, and fluidics
Application
Luminex assays
Protein assays/nucleic acid assays (ProcartaPlex / QuantiGene Plex)
Sample volume
6.3–80 µL (assay dependent)
25–80 µL (assay dependent)
Largest available panel
80
Plate format
96-well or 384-well
96-well or 384-well
96-well or 384-well
96-well
Software
21 CFR compliance
Yes
Automation Module
Yes
Yes
Integration software
Integration software
Analysis software
Data analysis apps for Luminex assays on the
Thermo Fisher Connect Platform
xPONENT basic plus Thermo Fisher Connect Platform
Although the MAGPIX instrument (RUO) has been discontinued we continue to offer all the accessories needed for this instrument.