M9 minimal medium (M9) is commonly used for the cultivation of bacteria (Miller, et al., 1972). It is comprised of a M9 minimal salts base formulation that can be supplemented with various amino acids and carbon sources. Life Technologies offers M9 minimal salts in a concentrated 2X ready-to-use liquid format for convenience and ease of use.
Ordering Information
Catalog # Name Size Price (USD) Qty
A1374401 M9 Minimal Salts (2X) Each
86.25