Microcentrifuge Tubes
Prepare, transfer and store samples securely
Whether you're preparing, transferring or storing samples, these multipurpose microcentrifuge tubes and caps can be used with confidence. Choose from a selection of tubes and caps that will suit your application.
Select the microcentrifuge tubes that's best for your application
Low DNA Binding Snap Cap
Microcentrifuge Tube
Snap Cap
Microcentrifuge Tube
Screw Cap
Microcentrifuge Tubes
Microtiter Tubes
Sterilin Certified
Universal
Short Term Cold Storage
Sample Preparation
Incubation & Water Bath
Short Term Cold Storage
Sample Preparation
Incubation & Water Bath
Short to Medium Term Storage
Sample Transportation
Sample Collection and Transformation
Serial Dilution
Sample Preparation
Plate Transfer
Sample Preparation
Sample Storage
Centrifugation
RNase, Dnase, Human and Mouse DNA free, non-Pyrogenic
Hydrophobic resin for maximum sample recovery
Low DNA binding
RNase, Dnase, Human and Mouse DNA free, non-Pyrogenic
Hydrophobic resin for maximum sample recovery
RNase, Dnase, Human and Mouse DNA free, non-Pyrogenic
Low Binding
96 Well Rack format
1.2ml volume level
RNase, DNase, human DNA free & non-Pyrogenic
Leak free 
Self-standing conical base
Lot number on container
Beveled cap for comfortable opening and closing
Available in multiple colors for easy sample identification
Beveled cap for comfortable opening and closing
Available in multiple colors for easy sample identification
Sample Recovery
Support automation filling and labeling systems
Provides Secure Seal
Support Plate Transfer
Additional tube volume for serial dilution and mixing
General microbiology and molecular applications
Larger volume sample management
0.6, 1.5, 2.0mL
Centrifugation Range: 25-30k x G
Graduation Marks
Non-Sterile
Autoclavable 120°C, 15PSI, 15 minutes
–80°C, –20°C, dry ice, wet ice
Biobased resin options
0.6, 1.5, 2.0mL
Centrifugation Range: 25-30k x G
Graduation Marks
Sterile & Non-Sterile
Autoclavable 120°C, 15PSI, 15 minutes
8 Different Colors: Amber, Red, Violet, Blue, Yellow, Green, Orange, Natural
–80°C, –20°C, dry ice, wet ice
0.5, 1.5, 2.0mL
Centrifuge Range:
18-25k x G
O-ring cap
Sterile & Non-Sterile
Skirted & Non Skirted
Knurled & Non Knurled
Colored Caps and Tethered Caps
–80°C, –20°C, dry ice, wet ice
1.2mL
Available 8 Strip Cap
Natural Only
Sterile & Non-Sterile
Autoclavable 120°C,15PSI,15 minutes
-80°C Storage
25mL
Plain or with graduations & marker spot
Aseptically manufactured and irradiated 
Maximum rcf 9,500 x g
-80oC storage
Learn more Order Order Order  Other
Microcentrifuge Tube Selection Tool
Find the screw cap microcentrifuge tube for your application. Select the tool to view and compare all available tubes.
1x1 image pixel for data collection