Search Thermo Fisher Scientific
As emerging infectious diseases and antimicrobial resistance shape the future of pharmaceutical and biotechnological development, molecular microbial detection has become commonplace in the workflow of various scientific processes. Microbial identification by real-time PCR (qPCR) offers a distinct advantage over culture-based methods, providing a highly specific means of detection without waiting on the growth of the organism. Contamination concerns in biopharmaceutical and biotechnology manufacturing processes highlight qPCR microbial identification assays as a solution for the microbial surveillance of various components of production.
What chromosome position are you interested in?
Genome Version: Build GRCh38 for
Instruments for real time PCR and digital PCR
Process and interpret qPCR data with intuitive software data analysis tools for a range of applications
Instruments for real time PCR and digital PCR
Process and interpret qPCR data with intuitive software data analysis tools for a range of applications
All assays employ Applied Biosystems TaqMan probe-based chemistries to produce highly sensitive and specific results with broad multiplexing capability.
Multiplex PCR is an effective way to help reduce the costs of and improve the efficiency of your microbial identification assays. The relative costs for a multiplex reaction are much closer to those for a singleplex reaction and offer substantial savings when compared to the costs of performing four singleplex reactions independently.
All TaqMan microbial identification assays are available with both FAM and VIC dye configurations accommodating the easy design of your duplex reactions. For higher order multiplexing with up to four targets, TaqMan microbial identification assays are also compatible with ABY and JUN dye configurations through Specialty Oligos.
For details and information around how best to design your multiplex assay to accommodate the needs of your laboratory, please see Multiplex Real-Time qPCR Solution.
For Research Use Only. Not for use in diagnostic procedures.