Search Thermo Fisher Scientific
Contains L-glutamine.
McCoy's Media were originally formulated for the propagation of human lymphocytes. McCoy's 5A medium (modified) was based on the original formulation of McCoy as modified by Hsu and Kellog and further modified by Iwakata and Grace.
Catalog Number(s)
Components | Molecular Weight | Concentration (mg/L) | mM |
---|---|---|---|
Amino Acids | |||
Glycine | 75.0 | 7.5 | 0.1 |
L-Alanine | 89.0 | 13.9 | 0.15617977 |
L-Arginine hydrochloride | 211.0 | 42.1 | 0.19952606 |
L-Asparagine | 132.0 | 45.0 | 0.3409091 |
L-Aspartic acid | 133.0 | 19.97 | 0.15015037 |
L-Cysteine | 121.0 | 31.5 | 0.2603306 |
L-Glutamic Acid | 147.0 | 22.1 | 0.15034014 |
L-Glutamine | 146.0 | 219.2 | 1.5013698 |
L-Histidine hydrochloride-H2O | 210.0 | 20.96 | 0.09980952 |
L-Hydroxyproline | 131.0 | 19.7 | 0.15038168 |
L-Isoleucine | 131.0 | 39.36 | 0.300458 |
L-Leucine | 131.0 | 39.36 | 0.300458 |
L-Lysine hydrochloride | 183.0 | 36.5 | 0.19945355 |
L-Methionine | 149.0 | 14.9 | 0.099999994 |
L-Phenylalanine | 165.0 | 16.5 | 0.1 |
L-Proline | 115.0 | 17.3 | 0.15043478 |
L-Serine | 105.0 | 26.3 | 0.25047618 |
L-Threonine | 119.0 | 17.9 | 0.15042016 |
L-Tryptophan | 204.0 | 3.1 | 0.0151960775 |
L-Tyrosine | 181.0 | 18.1 | 0.1 |
L-Valine | 117.0 | 17.6 | 0.15042736 |
Vitamins | |||
Ascorbic Acid | 176.0 | 0.5 | 0.0028409092 |
Biotin | 244.0 | 0.2 | 8.1967213E-4 |
Choline chloride | 140.0 | 5.0 | 0.035714287 |
D-Calcium pantothenate | 477.0 | 0.2 | 4.1928721E-4 |
Folic Acid | 441.0 | 10.0 | 0.022675738 |
Niacinamide | 122.0 | 0.5 | 0.0040983604 |
Nicotinic acid (Niacin) | 123.0 | 0.5 | 0.0040650405 |
Para-Aminobenzoic Acid | 137.0 | 1.0 | 0.00729927 |
Pyridoxal hydrochloride | 204.0 | 0.5 | 0.0024509805 |
Pyridoxine hydrochloride | 206.0 | 0.5 | 0.0024271845 |
Riboflavin | 376.0 | 0.2 | 5.319149E-4 |
Thiamine hydrochloride | 337.0 | 0.2 | 5.934718E-4 |
Vitamin B12 | 1355.0 | 2.0 | 0.0014760147 |
i-Inositol | 180.0 | 36.0 | 0.2 |
Inorganic Salts | |||
Calcium Chloride (CaCl2) (anhyd.) | 111.0 | 100.0 | 0.9009009 |
Magnesium Sulfate (MgSO4-7H2O) | 246.0 | 200.0 | 0.8130081 |
Potassium Chloride (KCl) | 75.0 | 400.0 | 5.3333335 |
Sodium Bicarbonate (NaHCO3) | 84.0 | 2200.0 | 26.190475 |
Sodium Chloride (NaCl) | 58.0 | 6460.0 | 111.37931 |
Sodium Phosphate monobasic (NaH2PO4-H2O) | 138.0 | 580.0 | 4.2028985 |
Other Components | |||
Bacto-Peptone | 600.0 | Infinity | |
D-Glucose (Dextrose) | 180.0 | 3000.0 | 16.666666 |
Glutathione (reduced) | 307.0 | 0.5 | 0.0016286644 |
Phenol Red | 376.4 | 10.0 | 0.026567481 |
1. Committee, In Vitro (1974) 9:6.
2. Hsu, T.C. and Kellogg, D.S., Jr. (1960) J. Nat. Cancer Inst., 25:221.
3. Iwakata, S. and Grace, J.T., Jr. (1964) N.Y.J. Med., 64:2279.
4. McCoy, T.A., Maxwell, M. and Kruse, P.F. (1959) Proc. Soc. Exper. Biol. Med., 100:115.
5. Sertich, G.J., Glass, J.R., Fuller, D.J.M., et al., (1986) J. Cell. Physiol., 127:114. (CHO adherent cells).
16600 - McCoy's 5A (modified)
Cat No:
16600082
,
16600108
12330 - McCoy's 5A (modified), HEPES
Cat No:
12330031
80014 - McCoy's 5A (Park and Terasaki modification)
Cat No:
80014020