Adult rat cortical astrocyte. Rhod-2 AM and MitoFluor™ Green stain.
Colocalization of fluorescent staining by rhod-2 AM (Cat. no. R1244, R1245MP; upper panel) and mitochondrion-selective MitoFluor™ Green stain (Cat. no. M7502; lower panel) in an adult rat cortical astrocyte. Cells were simultaneously loaded with rhod-2 AM (4.5 µM) and the MitoFluor™ Green stain (20 nM) for 30 minutes at 22°C. Confocal laser-scanning microscopy using 488 nm excitation and spectrally resolved detection, at 505–530 nm for MitoFluor™ Green stain and >=585 nm for rhod-2, shows almost identical staining distribution. The images were contributed by Michael Duchen, University College, London. Reproduced with permission from J Cell Biol 145, 795 (1999).
Colocalization of fluorescent staining by rhod-2 AM (Cat. no. R1244, R1245MP; upper panel) and mitochondrion-selective MitoFluor™ Green stain (Cat. no. M7502; lower panel) in an adult rat cortical astrocyte. Cells were simultaneously loaded with rhod-2 AM (4.5 µM) and the MitoFluor™ Green stain (20 nM) for 30 minutes at 22°C. Confocal laser-scanning microscopy using 488 nm excitation and spectrally resolved detection, at 505–530 nm for MitoFluor™ Green stain and >=585 nm for rhod-2, shows almost identical staining distribution.  The images were contributed by Michael Duchen, University College, London. Reproduced with permission from J Cell Biol 145, 795 (1999).
Related Products
Catalog # Name Size Price (EUR) Qty
R1244 Rhod-2, AM, cell permeant, 1mg Each
808,00
R1245MP Rhod-2, AM, cell permeant, 20 x 50μg Each
928,00
Related Images