Thermo Fisher Scientific  ›  Electron Microscopy   Electron Microscopes     3D Visualization, Analysis and EM Software   Customer Portal
On-demand webinar: DVC in Avizo Software for material deformation analysis
The Thermo Scientific Avizo Software is a comprehensive digital lab for image analysis. It allows you to measure and analyze 3D displacement and strain maps from volume images taken before and after material deformation. The software not only visualizes the changes but also helps quantify the microstructural changes caused by deformation, such as strain transfer in materials with multiple phases, pore growth and coalescence related to fractures, and crack opening displacement.
Watch or re-watch this webinar and learn:
  • How to design a DVC experiment
  • The difference between global and local approaches to DVC
  • How DVC is used with large datasets 
  • The advantage of mechanical regularization
  • How DVC can help inform and validate numerical simulations
>>Watch it now >