11 Oct 2019
Berea Pore Size Distribution Tutorial
Create a semi-automatic workflow for Pore Size Distribution on Berea sandstone.
An example of Pore Size Distribution computation on Cubic
Micro-CT Berea Sandstone.
The workflow is split into two recipes for easier
utilization with other datasets.
First recipe focuses on filtering and segmenting the pore
space. The second recipe splits the pore space into individual pores, computes
length and volume for each pore and finally computes the Pore Size
Distribution.