15 Apr 2020
Copy Measure Annotation Settings
This Tcl script-object can be used to copy visualisation settings from one measure to all others in a Measurement module. It is also possible to save/load settings to/from a text file.
![CopyMeasureAnnotationSettings2 CopyMeasureAnnotationSettings2](/software-em-3d-vis/xtra-library/uploads/2020/01/CopyMeasureAnnotationSettings2-lg.png)
The following visualisation settings
are considered:
calloutFontTitle calloutFontNote calloutColors decimalPlaces lineWidth color
Right-click on a Measurement module, select Create Object and choose Xtra->Annotate->CopyMeasureAnnotationSettings.
The parameters are:
- Reference measure: Use the slider to select the measure whose settings shall be copied to all the others. The label of the measure is shown below in Measure name.
- filename: You can specify a file name here, for saving or retrieving settings.
- Action:
- Import settings: After importing settings, those settings will be used when pressing Apply, as long as no other things were changed afterwards (e.g. the Reference measure index selection).
- Export settings: The settings of the selected measure will be saved to the specified file.
Press Apply to apply the chosen settings to all measures of the Measurement object.