Efficient Graph-based 2D Image Segmentation
This Xtra implements efficient graph-based image segmentation by Felzenszwalb and Huttenlocher.
![2021-01-15 12_39_25-Amira - GraphBasedSegmentation_volumescope.hx 2021-01-15 12_39_25-Amira - GraphBasedSegmentation_volumescope.hx](/software-em-3d-vis/xtra-library/uploads/2021/01/2021-01-15 12_39_25-Amira - GraphBasedSegmentation_volumescope.hx-lg.png)
This Xtra implements efficient graph-based image segmentation by Felzenszwalb and Huttenlocher [1].
The new GraphBasedSegmentation module is stored in the category Xtra>Image Processing. It performs 2D, XY slice-by-slice segmentation of the input grayscale image. You can use this form of segmentation to perform over-segmentation of grayscale images.
The project included in this Xtra demonstrates quick partitioning and labeling of a VolumeScope SEM image of a mouse brain to isolate objects that are circular or elliptical and that have relatively higher brightness compared to the other objects in the scene.
[1] Felzenszwalb, Pedro F., and Daniel P. Huttenlocher. "Efficient graph-based image segmentation." International Journal of Computer Vision (IJCV), 2004.
Download a version compatible with Amira-Avizo 2022.1 HERE, 2022.2 HERE, 2023.1 HERE.
To obtain a version of this Xtra compatible with Amira-Avizo 2024.1 and further, please contact us via our Customer Support Center.