28 Apr 2022
Extract Color Wash Image
This module exports an image of color-washed data.
The Extract Color Wash Image module exports the data displayed by a Color Wash module into a new data field. The exported field is a one slice color field (HxUniformColorField3) that can be saved as an image file by the "Export Data As..." command.
This module requires a Color Wash module as input that is connected to an Ortho Slice module.