11 May 2020
Extract Label
This module extracts a specified label region by its ID.
![snapshot-extract-label-2 snapshot-extract-label-2](/software-em-3d-vis/xtra-library/uploads/2020/02/snapshot-extract-label-2-lg.png)
Description:
Extract a specified label region by the ID from a label field. The module supports 8, 16 and 32 bit labels.
Connections:
- Labels [required]: A label field (HxUniformLabelField3) to be extracted.
Ports:
- Label ID [required]: A label ID to be extracted. The parameter range is automatically adjusted by the connected label field.
- Export as binary: If checked
'yes', a binary result is exported. This option is useful for subsequent
computation (e.g. for Separate Objects module). Otherwise, the original label
value is retained. It is useful for visualization with same colormap of the
input image for this module.