How to Use the Image Recipe Designer Workroom
This tutorial demonstrates how to use the Image Recipe Designer (formerly 'Image Stack Processing' or 'ISP') workroom, an intuitive 2D workroom for automating image processing workflows in 3D.
Important note: this tutorial has been recorded with Amira-Avizo 2020.2. It demonstrates how to use the Image Stack Processing (ISP) workroom. Amira-Avizo 2022.2 release introduced some changes in the image recipes creation and edition management. To facilitate user's experience, the 'Image Stack Processing' (ISP) and 'Image Volume Processing' (IVP) features have been merged into one feature: 'Image Recipe'. The new workroom is now called 'Image Recipe Designer' (previously ISP or IVP workrooms). The new module for executing a recipe on a dataset in the Project View is called 'Image Recipe Player' (previously 'Image Stack Processing' or 'Image Volume Processing'). The philosophy of the workroom remains the same, therefore this tutorial remains fully relevant even if you are using the latest version of Amira-Avizo Software.
This tutorial demonstrates, using a simple example, how to use the Image Stack Processing (ISP) workroom. A simple workflow composed of basic filtering and thresholding operations is performed on the dataset by first tuning it in 2D in the ISP workroom, and then applying it to the complete 2D stack from the main Project workroom.
Get a tour of the ISP workroom
The Image Stack Processing module and workroom allow for the creation and execution of image processing workflows to process an image stack in 2D (each image of the stack is processed individually). The Image Stack Processing editing workroom features an interactive and dynamic edition environment, giving constant feedback about the workflow being built. Any step of the recipe can be accessed and modified, with immediate feedback about the impact of each change on the final output. It is an optimized 2D setup that enhances and segments datasets in the Amira-Avizo software.