30 Dec 2021
How to Use the Surface View Module and Create Elaborate Surface Renderings
This short video demonstrates the basic use of the Surface View module, and how to combine several Surface View modules to create elaborate renderings.
This tutorial introduces the basic use of the Surface View module to display a surface in the following ways:
- Modify the Draw Style options.
- Use the Buffer to select the materials to be displayed or hidden.
- Use a second Surface View module and play with its transparency to create an in-depth rendering of the materials composing the surface.
Additional resources: