22 Jan 2021
Point Cloud to Spreadsheet
This Python script-object allows you to convert a point-cloud object to a spreadsheet object.
PointCloudToSpreadsheet
This Python script-object converts a point-cloud object into a spreadsheet-object. This can be useful, for example, if you want to:
  • Export the points' coordinates to some other software, because spreadsheets can be exported to ASCII, CSV, or XML formats.
  • Perform some statistical analysis on the points' coordinates.
The point coordinates are written to the columns CenterX, CenterY, and CenterZ. If data-columns exist in the point-cloud object, those are also copied to the spreadsheet.
Connect the script-object to the point-cloud object and click Apply.
In the example project, the local density of a point cloud is calculated and assigned to the points. To get a table out of this, Point Cloud To Spreadsheet is used.
Note: This feature is now natively included in the Amira-Avizo Software, starting from version 2024.1.