24 Feb 2022
Slab View
The Slab View display module allows you to display only a slab from any kind of 3D visualization.
The Slab View display module is basically a combination of a slice and a clipping plane, synchronized in position and orientation, and with a user-defined offset. This allows the clipping plane to remain at a fixed distance when moving the slice.
Clipping can be activated for the slice, the clipping plane, or both, in the same or in the opposite direction, so that it is possible to display only the slab between the slice and the clipping plane.
Also, the slice can be rotated interactively and the clipping plane can be realigned onto it afterwards.
The visibility of the Slab View can be toggled independently for each viewer.