Split and Merge Segments
The Split and Merge Segments module merges the two most representative segments at the intersection points of a graph structure and detaches the others.
Currently the segmentation of individual branches of a tree structure (vascular system for instance) is not an easy task and requires a lot of manual editing. This Xtra provides a solution to make an almost fully automatic workflow to achieve this individual segmentation of the branches.
The Split and Merge Segments module merges, at each branching point of the graph, the two segments with the highest evaluation score, and detaches the other segments from this branching point.
For each detached segment, a new node is created as the one new end of this segment.
The evaluation score is computed from one of the point attributes available.
Once a point attribute has been selected for the evaluation, the evaluation scoring and ranking methods can be defined.
For example, if you pick the 'thickness' point attribute for the evaluation, 'mean' for the scoring, and 'biggest' for the ranking, then the two segments with the biggest thickness are merged at each intersection point of the graph, and the other segments are detached.
Download a version compatible with Amira-Avizo 2022.1 HERE, 2022.2 HERE, 2023.1 HERE.
To obtain a version of this Xtra compatible with Amira-Avizo 2024.1 and further, please contact us via our Customer Support Center.