Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAPCP-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101496.3 | NP_173080.1 | 1 - 2 | 267 | 98 | |
GenBank mRNA | BT021096.1 | - | 1 - 2 | 147 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |