Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SR45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198085.1 | NP_001185014.1 | 3 - 4 | 521 | 77 | |
NM_001332238.1 | NP_001321127.1 | 3 - 4 | 510 | 77 | ||
NM_001332239.1 | NP_001321128.1 | 3 - 4 | 542 | 77 | ||
NM_001332240.1 | NP_001321125.1 | 3 - 4 | 525 | 77 | ||
NM_001332241.1 | NP_001321126.1 | 3 - 4 | 525 | 77 | ||
NM_101523.4 | NP_173107.1 | 3 - 4 | 510 | 77 | ||
NM_202115.3 | NP_973844.1 | 3 - 4 | 510 | 77 | ||
GenBank mRNA | AF151366.1 | - | 3 - 4 | 392 | 77 | |
BT020372.1 | - | 3 - 4 | 355 | 77 | ||
BT021095.1 | - | 3 - 4 | 375 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |