Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332389.1 | NP_001320535.1 | 5 - 6 | 962 | 153 | |
NM_101785.4 | NP_173361.1 | 4 - 5 | 1155 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |