Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMGB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035997.1 | NP_001031074.1 | 5 - 6 | 438 | 69 | |
NM_001084100.1 | NP_001077569.1 | 5 - 6 | 438 | 69 | ||
NM_101920.3 | NP_564123.1 | 5 - 6 | 466 | 69 | ||
GenBank mRNA | AB493468.1 | - | 4 - 5 | 252 | 69 | |
AF370248.1 | - | 5 - 6 | 435 | 69 | ||
AK118543.1 | - | 5 - 6 | 350 | 69 | ||
AY063064.1 | - | 4 - 5 | 252 | 69 | ||
Y14072.1 | - | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |