Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STR16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126001.4 | NP_569026.2 | 2 - 3 | 465 | 61 | |
NM_180947.3 | NP_851278.1 | 3 - 4 | 505 | 61 | ||
GenBank mRNA | AY049302.1 | - | 3 - 4 | 329 | 61 | |
AY084763.1 | - | 3 - 4 | 320 | 61 | ||
BT000864.1 | - | 3 - 4 | 292 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |