Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ATMAP65-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035868.2 | NP_001030945.1 | 8 - 9 | 1307 | 77 | |
NM_126252.4 | NP_178300.3 | 8 - 9 | 2494 | 77 | ||
GenBank mRNA | AK317525.1 | - | 8 - 9 | 1605 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |