Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_126341.3 | NP_178389.1 | 6 - 7 | 933 | 61 | |
NM_201675.2 | NP_973404.1 | 6 - 7 | 908 | 61 | ||
GenBank mRNA | AJ289165.1 | - | 6 - 7 | 610 | 61 | |
AK226970.1 | - | 6 - 7 | 737 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |