Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111016.4 | NP_186799.2 | 9 - 10 | 1290 | 110 | |
NM_180159.2 | NP_850490.1 | 9 - 10 | 1102 | 110 | ||
GenBank mRNA | AF428284.1 | - | 9 - 10 | 1054 | 110 | |
AF428459.1 | - | 9 - 10 | 1104 | 110 | ||
AY091066.1 | - | 9 - 10 | 1099 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |