Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTAC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036597.2 | NP_001031674.1 | 1 - 2 | 560 | 78 | |
NM_118120.5 | NP_567593.1 | 1 - 2 | 608 | 78 | ||
GenBank mRNA | BT026461.1 | - | 1 - 2 | 415 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |