Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: QQT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_118300.3 | NP_193911.1 | 5 - 6 | 753 | 90 | |
NM_202857.2 | NP_974586.1 | 5 - 6 | 848 | 90 | ||
GenBank mRNA | AF419563.1 | - | 5 - 6 | 753 | 90 | |
AY143879.1 | - | 4 - 5 | 669 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |