Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBP27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342538.1 | NP_001320167.1 | 6 - 7 | 1351 | 138 | |
NM_120097.3 | NP_568058.1 | 7 - 8 | 1271 | 138 | ||
GenBank mRNA | AF302675.1 | - | 7 - 8 | 1101 | 138 | |
AF370208.1 | - | 7 - 8 | 1236 | 138 | ||
AY133770.1 | - | 7 - 8 | 1083 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |