Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEE13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161045.2 | NP_001154517.1 | 3 - 4 | 317 | 77 | |
NM_001202597.1 | NP_001189526.1 | 3 - 4 | 317 | 77 | ||
NM_001335430.1 | NP_001324683.1 | 3 - 4 | 317 | 77 | ||
NM_001335431.1 | NP_001324684.1 | 3 - 4 | 317 | 77 | ||
NM_001335432.1 | NP_001324685.1 | 2 - 3 | 447 | 77 | ||
NM_001335433.1 | NP_001324686.1 | 2 - 3 | 405 | 77 | ||
NM_127032.2 | NP_179075.1 | 2 - 3 | 447 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |