Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204043.2 | NP_001190972.1 | 7 - 8 | 1777 | 142 | |
NM_001204044.1 | NP_001190973.1 | 7 - 8 | 1774 | 142 | ||
NM_001342558.1 | NP_001328232.1 | 6 - 7 | 2141 | 142 | ||
NM_120148.3 | NP_568072.1 | 7 - 8 | 1764 | 142 | ||
GenBank mRNA | AF378120.1 | - | 7 - 8 | 1693 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |