Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: FRK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127476.2 | NP_179509.1 | 5 - 6 | 1384 | 90 | |
GenBank mRNA | AF486619.1 | - | 5 - 6 | 1384 | 90 | |
FJ708695.1 | - | 5 - 6 | 1333 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |