Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ckl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127505.5 | NP_179537.1 | 6 - 7 | 813 | 118 | |
GenBank mRNA | AY086262.1 | - | 6 - 7 | 745 | 118 | |
AY099837.1 | - | 6 - 7 | 704 | 118 | ||
AY943844.1 | - | 6 - 7 | 506 | 118 | ||
BT006580.1 | - | 6 - 7 | 506 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |