Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: DPB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203285.1 | NP_001190214.1 | 9 - 10 | 1269 | 125 | |
NM_180426.3 | NP_850757.1 | 9 - 10 | 1088 | 125 | ||
GenBank mRNA | AB493733.1 | - | 8 - 9 | 828 | 125 | |
AJ294532.1 | - | 8 - 9 | 828 | 125 | ||
AK228148.1 | - | 9 - 10 | 1069 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |