Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BON2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120812.4 | NP_568180.1 | 6 - 7 | 912 | 136 | |
GenBank mRNA | AF389301.1 | - | 6 - 7 | 676 | 136 | |
BT010130.1 | - | 6 - 7 | 641 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |