Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BIM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125718.1 | NP_001119190.1 | 5 - 6 | 1324 | 150 | |
NM_001203328.2 | NP_001190257.1 | 6 - 7 | 1271 | 150 | ||
NM_001203329.1 | NP_001190258.1 | 6 - 7 | 1271 | 150 | ||
NM_001203330.2 | NP_001190259.1 | 6 - 7 | 1283 | 150 | ||
NM_001342983.1 | NP_001318507.1 | 6 - 7 | 1107 | 150 | ||
NM_001342984.1 | NP_001318508.1 | 6 - 7 | 1107 | 150 | ||
NM_001342985.1 | NP_001330723.1 | 6 - 7 | 1271 | 150 | ||
NM_001342986.1 | NP_001330724.1 | 6 - 7 | 1283 | 150 | ||
GenBank mRNA | AB493742.1 | - | 3 - 4 | 412 | 150 | |
BT029780.1 | - | 3 - 4 | 412 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |