Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: TGG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122501.3 | NP_197972.2 | 6 - 7 | 824 | 120 | |
NM_180746.3 | NP_851077.1 | 6 - 7 | 824 | 120 | ||
GenBank mRNA | AY045681.1 | - | 6 - 7 | 818 | 120 | |
AY090382.1 | - | 6 - 7 | 787 | 120 | ||
L11454.1 | - | 6 - 7 | 820 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |