Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KAT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036960.2 | NP_001032037.1 | 10 - 11 | 935 | 80 | |
NM_001344808.1 | NP_001331840.1 | 10 - 11 | 1107 | 80 | ||
NM_124265.5 | NP_568704.2 | 9 - 10 | 1802 | 80 | ||
NM_180826.4 | NP_851157.1 | 9 - 10 | 952 | 80 | ||
GenBank mRNA | AF062590.1 | - | 9 - 10 | 832 | 80 | |
AF062591.1 | - | 9 - 10 | 1051 | 80 | ||
AY079026.1 | - | 9 - 10 | 937 | 80 | ||
AY136455.1 | - | 9 - 10 | 840 | 80 | ||
BT002594.1 | - | 8 - 9 | 739 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |