Search Thermo Fisher Scientific
Gene Symbol: FIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_124882.4 | NP_200311.1 | 6 - 7 | 609 | 91 | |
NM_180862.1 | NP_851193.1 | 6 - 7 | 604 | 91 | ||
GenBank mRNA | AF174429.1 | - | 6 - 7 | 604 | 91 | |
BT030093.1 | - | 5 - 6 | 523 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |