Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345242.1 | NP_001331281.1 | 11 - 12 | 1172 | 92 | |
NM_125093.6 | NP_200521.3 | 11 - 12 | 1221 | 92 | ||
NM_180869.3 | NP_851200.1 | 11 - 12 | 1063 | 92 | ||
GenBank mRNA | AJ249352.1 | - | 11 - 12 | 1065 | 92 | |
AK226961.1 | - | 10 - 11 | 987 | 92 | ||
AY069869.1 | - | 11 - 12 | 1065 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |