Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035765.2 | NP_001030842.1 | 9 - 10 | 921 | 75 | |
NM_001339563.1 | NP_001326925.1 | 9 - 10 | 885 | 75 | ||
NM_001339564.1 | NP_001326924.1 | 6 - 7 | 518 | 75 | ||
NM_115078.4 | NP_566960.1 | 9 - 10 | 1017 | 75 | ||
GenBank mRNA | AJ302781.1 | - | 9 - 10 | 802 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |