Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: MDAR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161202.1 | NP_001154674.1 | 6 - 7 | 557 | 150 | |
NM_115148.4 | NP_190856.1 | 5 - 6 | 604 | 150 | ||
GenBank mRNA | AF360197.1 | - | 5 - 6 | 399 | 150 | |
AK227030.1 | - | 5 - 6 | 428 | 150 | ||
AK317341.1 | - | 5 - 6 | 427 | 150 | ||
AY045666.1 | - | 5 - 6 | 427 | 150 | ||
AY057576.1 | - | 5 - 6 | 455 | 150 | ||
AY060525.1 | - | 5 - 6 | 373 | 150 | ||
AY070718.1 | - | 5 - 6 | 446 | 150 | ||
AY091403.1 | - | 5 - 6 | 373 | 150 | ||
AY125492.1 | - | 5 - 6 | 425 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |