Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_103495.4 | NP_564486.1 | 1 - 2 | 711 | 66 | |
GenBank mRNA | AY065453.1 | - | 1 - 2 | 664 | 66 | |
AY117284.1 | - | 1 - 2 | 590 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |