Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BGAL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333243.1 | NP_001320473.1 | 4 - 5 | 492 | 113 | |
NM_103587.3 | NP_175127.1 | 6 - 7 | 1024 | 113 | ||
GenBank mRNA | AY069911.1 | - | 6 - 7 | 823 | 113 | |
AY093977.1 | - | 6 - 7 | 688 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |