Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDA14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115900.4 | NP_191595.1 | 6 - 7 | 629 | 75 | |
GenBank mRNA | AY039876.1 | - | 6 - 7 | 601 | 75 | |
AY085156.1 | - | 6 - 7 | 584 | 75 | ||
AY101514.1 | - | 5 - 6 | 410 | 75 | ||
BT000725.1 | - | 6 - 7 | 545 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |