Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PBF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035828.2 | NP_001030905.1 | 2 - 3 | 287 | 97 | |
NM_001203217.1 | NP_001190146.1 | 2 - 3 | 319 | 97 | ||
NM_115946.3 | NP_191641.1 | 2 - 3 | 287 | 97 | ||
GenBank mRNA | AF043537.1 | - | 2 - 3 | 242 | 97 | |
AY062790.1 | - | 2 - 3 | 256 | 97 | ||
AY081571.1 | - | 2 - 3 | 157 | 97 | ||
AY086619.1 | - | 2 - 3 | 277 | 97 | ||
X79806.1 | - | 216 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |