Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AK-LYS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121331.3 | NP_196832.1 | 8 - 9 | 1365 | 86 | |
GenBank mRNA | AY057674.1 | - | 8 - 9 | 1239 | 86 | |
BT000493.1 | - | 8 - 9 | 1161 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |