Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BBM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343497.1 | NP_001332647.1 | 7 - 8 | 1011 | 63 | |
NM_121749.3 | NP_197245.2 | 8 - 9 | 1209 | 63 | ||
GenBank mRNA | EF687843.1 | - | 8 - 9 | 1029 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |